Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037585.1 | 1 | 91 | |||
NR_037586.1 | 1 | 91 | ||||
GenBank mRNA | BC130500.1 | - | 1 | 62 | ||
BC130504.1 | - | 1 | 62 | |||
BC144390.1 | - | 1 | 62 | |||
BC144391.1 | - | 1 | 62 | 9052912 | ||
BX089409.1 | - | 1 | 90 | |||
N47348.1 | - | 1 | 91 |