Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEMIS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010923.2 | NP_001010923.1 | 4 | 1686 | ||
NM_001164685.1 | NP_001158157.1 | 4 | 1686 | |||
NM_001164687.1 | NP_001158159.1 | 4 | 1650 | |||
NM_001318531.1 | NP_001305460.1 | 5 | 1751 | |||
XM_011535813.2 | XP_011534115.1 | 6 | 1880 | |||
XM_011535814.2 | XP_011534116.1 | 5 | 1651 | |||
XM_011535816.1 | XP_011534118.1 | 6 | 1876 | |||
XM_017010848.1 | XP_016866337.1 | 6 | 1880 | |||
XM_017010849.1 | XP_016866338.1 | 4 | 1394 | |||
GenBank mRNA | AK097903.1 | - | 4 | 1663 | ||
AK124031.1 | - | 4 | 1616 | |||
AK128377.1 | - | 5 | 1684 | |||
AK292046.1 | - | 4 | 1636 | |||
BC130516.1 | - | 4 | 1680 | 40146547 | ||
BC144034.1 | - | 4 | 1678 |