Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM208B SIRNA Assays ›
Gene Symbol:
FAM208B
Gene Name:
family with sequence similarity 208 member B
Gene Aliases:
C10orf18, bA318E3.2
Chromosome Location:
Chr.10: 5684759 - 5763740 on Build GRCh38
UniGene ID:
Hs.610717
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
25543
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM208B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321783.1
NP_001308712.1
16
5932
NM_001321784.1
NP_001308713.1
16
5978
NM_001321785.1
NP_001308714.1
15
5689
NM_017782.4
NP_060252.4
15
5859
XM_005252474.3
XP_005252531.1
18
12670
XM_005252475.3
XP_005252532.1
19
6331
XM_005252476.4
XP_005252533.1
17
6114
XM_005252480.4
XP_005252537.1
15
5919
XM_005252481.4
XP_005252538.1
13
5404
XM_005252482.2
XP_005252539.1
17
5764
XM_011519515.2
XP_011517817.1
19
12808
XM_011519516.2
XP_011517818.1
19
12808
XM_011519517.2
XP_011517819.1
19
6728
XM_011519518.2
XP_011517820.1
18
6041
XM_011519519.2
XP_011517821.1
18
12565
XM_011519520.2
XP_011517822.1
16
12505
XM_011519521.2
XP_011517823.1
18
6252
XM_011519522.2
XP_011517824.1
18
6636
XM_011519525.2
XP_011517827.1
16
5672
XM_017016361.1
XP_016871850.1
18
12565
XM_017016362.1
XP_016871851.1
17
6535
XM_017016363.1
XP_016871852.1
18
6193
XM_017016364.1
XP_016871853.1
16
6397
XM_017016365.1
XP_016871854.1
17
6059
GenBank mRNA
AB095927.1
-
2
2047
AK000367.1
-
1
246
AK128051.1
-
3
3134
AK128055.1
-
1
1554
AK128151.1
-
1
1079
AK225352.1
-
1
246
AL832762.1
-
2
2562
BC001759.1
-
1
252
BC071976.1
-
1
280
BX649177.1
-
7
4832
CR936780.1
-
15
5860
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0