Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304746.1 | NP_001291675.1 | 2 | 384 | ||
NM_017821.4 | NP_060291.2 | 2 | 414 | |||
GenBank mRNA | AK097621.1 | - | 2 | 1149 | ||
AK294115.1 | - | 2 | 384 | |||
AY126343.1 | - | 1 | 140 | |||
BC013103.1 | - | 2 | 385 | |||
BC137108.1 | - | 1 | 263 | 9021105 | ||
BC137110.1 | - | 1 | 263 | 9021107 | ||
BC144476.1 | - | 1 | 263 | |||
BX951305.1 | - | 2 | 369 | |||
DB036310.1 | - | 2 | 414 |