Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014242.1 | NP_001014264.1 | 5 | 1830 | ||
GenBank mRNA | AB108669.1 | - | 5 | 1830 |
Gene Symbol: LOC103694869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773818.1 | XP_008772040.1 | 5 | 1849 |