Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CT55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031705.2 | NP_001026875.1 | 2 | 784 | ||
NM_017863.2 | NP_060333.1 | 2 | 784 | |||
XM_005262427.1 | XP_005262484.1 | 2 | 355 | |||
GenBank mRNA | AF520814.2 | - | 2 | 784 | ||
AK000534.1 | - | 2 | 291 | |||
AY352211.1 | - | 2 | 784 | |||
BC020662.1 | - | 2 | 193 | 4809793 |