Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281932.1 | NP_001268861.1 | 17 | 2069 | ||
NM_001281933.1 | NP_001268862.1 | 16 | 2016 | |||
NM_017760.6 | NP_060230.5 | 16 | 2016 | |||
NR_104054.1 | 19 | 2227 | ||||
XM_005249547.2 | XP_005249604.1 | 16 | 2014 | |||
XM_011516356.2 | XP_011514658.1 | 16 | 2014 | |||
XM_011516357.2 | XP_011514659.1 | 17 | 2042 | |||
XM_011516358.2 | XP_011514660.1 | 16 | 2014 | |||
XM_011516362.1 | XP_011514664.1 | 4 | 493 | |||
XM_017012374.1 | XP_016867863.1 | 16 | 2014 | |||
XM_017012377.1 | XP_016867866.1 | 6 | 772 | |||
GenBank mRNA | AK000318.1 | - | 11 | 1274 | ||
AK022744.1 | - | 6 | 816 | |||
AK023347.1 | - | 5 | 570 | |||
AK092008.1 | - | 17 | 2004 | |||
AK301901.1 | - | 4 | 494 | |||
AK315699.1 | - | 14 | 1633 | |||
BC020560.1 | - | 11 | 1340 | 3458938 | ||
BC043404.1 | - | 16 | 1957 | 6045248 | ||
BX537845.1 | - | 16 | 1983 |