Search Thermo Fisher Scientific
Gene Symbol: LRRC40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017768.4 | NP_060238.3 | 15 | 2688 | ||
XM_005271013.1 | XP_005271070.1 | 14 | 2515 | |||
XM_011541763.1 | XP_011540065.1 | 13 | 2430 | |||
XM_017001695.1 | XP_016857184.1 | 14 | 2436 | |||
XM_017001696.1 | XP_016857185.1 | 12 | 2210 | |||
GenBank mRNA | AK022951.1 | - | 15 | 2602 | ||
AL162076.1 | - | 5 | 1263 | |||
BC003407.1 | - | 15 | 2488 | |||
BC008586.1 | - | 15 | 2586 | 2900193 |