Search
Search

LFNG| Gene Symbol: LFNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040167.1 | NP_001035257.1 | 8 | 2275 | ||
| NM_001166355.1 | NP_001159827.1 | 9 | 2171 | |||
| NM_002304.2 | NP_002295.1 | 9 | 1959 | |||
| GenBank mRNA | AK096284.1 | - | 9 | 2171 | ||
| BC014851.1 | - | 9 | 1965 | 4453156 | ||