Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKRD36 SIRNA Assays ›
Gene Symbol:
ANKRD36
ANKRD36B
Gene Name:
ankyrin repeat domain 36
ankyrin repeat domain 36B
Gene Aliases:
UNQ2430
KIAA1641
Chromosome Location:
Chr.2: 97113186 - 97264521 on Build GRCh38
Chromosome Location:
Chr.2: 97492663 - 97590245 on Build GRCh38
UniGene ID:
Hs.541894
Hs.532921
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
256726
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKRD36
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164315.1
NP_001157787.1
3
632
XM_006712514.3
XP_006712577.1
3
942
XM_011511130.1
XP_011509432.1
3
942
XM_011511131.1
XP_011509433.1
3
942
XM_017004009.1
XP_016859498.1
3
942
XM_017004010.1
XP_016859499.1
3
942
XM_017004011.1
XP_016859500.1
3
942
XM_017004012.1
XP_016859501.1
3
942
XM_017004013.1
XP_016859502.1
3
942
XM_017004014.1
XP_016859503.1
3
942
XM_017004015.1
XP_016859504.1
3
942
XM_017004016.1
XP_016859505.1
3
942
XM_017004017.1
XP_016859506.1
3
942
XM_017004018.1
XP_016859507.1
3
942
XM_017004019.1
XP_016859508.1
3
942
XM_017004020.1
XP_016859509.1
3
942
XM_017004021.1
XP_016859510.1
3
942
XM_017004022.1
XP_016859511.1
3
942
XM_017004023.1
XP_016859512.1
3
942
XM_017004024.1
XP_016859513.1
3
942
XM_017004025.1
XP_016859514.1
3
942
XM_017004026.1
XP_016859515.1
3
942
XM_017004027.1
XP_016859516.1
3
942
XM_017004028.1
XP_016859517.1
3
942
XM_017004029.1
XP_016859518.1
3
942
XM_017004030.1
XP_016859519.1
3
942
XM_017004031.1
XP_016859520.1
3
942
XM_017004032.1
XP_016859521.1
3
942
XM_017004033.1
XP_016859522.1
3
942
XM_017004034.1
XP_016859523.1
3
942
XM_017004035.1
XP_016859524.1
3
942
XM_017004036.1
XP_016859525.1
3
942
XM_017004037.1
XP_016859526.1
3
942
XM_017004038.1
XP_016859527.1
3
942
XM_017004039.1
XP_016859528.1
3
942
XM_017004040.1
XP_016859529.1
3
942
XM_017004041.1
XP_016859530.1
3
942
XM_017004042.1
XP_016859531.1
3
942
XM_017004043.1
XP_016859532.1
3
942
XM_017004044.1
XP_016859533.1
3
942
XM_017004045.1
XP_016859534.1
3
942
XM_017004046.1
XP_016859535.1
3
942
XM_017004047.1
XP_016859536.1
3
942
XM_017004048.1
XP_016859537.1
3
942
XM_017004049.1
XP_016859538.1
3
942
GenBank mRNA
AK304740.1
-
3
572
AK307629.1
-
3
539
AK316541.1
-
3
572
AY358571.1
-
3
631
BC046186.1
-
3
511
BC128262.1
-
3
471
BC128517.1
-
3
471
40118468
BX647769.1
-
3
577
Gene Symbol: ANKRD36B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
AK074570.1
-
3
533
BM511319.1
-
3
224
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0