Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKRD36 SIRNA Assays ›
Gene Symbol:
ANKRD36
ANKRD36C
Gene Name:
ankyrin repeat domain 36
ankyrin repeat domain 36C
Gene Aliases:
UNQ2430
-
Chromosome Location:
Chr.2: 97113186 - 97264521 on Build GRCh38
Chromosome Location:
Chr.2: 95849548 - 95992320 on Build GRCh38
UniGene ID:
Hs.541894
Hs.646318
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
256727
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKRD36
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164315.1
NP_001157787.1
15
1455
XM_006712514.3
XP_006712577.1
15
1765
XM_011511130.1
XP_011509432.1
15
1765
XM_011511131.1
XP_011509433.1
15
1765
XM_017004009.1
XP_016859498.1
15
1765
XM_017004010.1
XP_016859499.1
15
1765
XM_017004011.1
XP_016859500.1
15
1765
XM_017004012.1
XP_016859501.1
15
1765
XM_017004013.1
XP_016859502.1
15
1765
XM_017004014.1
XP_016859503.1
15
1765
XM_017004015.1
XP_016859504.1
13
1663
XM_017004016.1
XP_016859505.1
15
1765
XM_017004017.1
XP_016859506.1
15
1765
XM_017004018.1
XP_016859507.1
15
1765
XM_017004019.1
XP_016859508.1
15
1765
XM_017004020.1
XP_016859509.1
15
1765
XM_017004021.1
XP_016859510.1
15
1765
XM_017004022.1
XP_016859511.1
15
1765
XM_017004023.1
XP_016859512.1
15
1765
XM_017004024.1
XP_016859513.1
14
1627
XM_017004025.1
XP_016859514.1
15
1765
XM_017004026.1
XP_016859515.1
15
1765
XM_017004027.1
XP_016859516.1
15
1765
XM_017004028.1
XP_016859517.1
15
1765
XM_017004029.1
XP_016859518.1
15
1765
XM_017004030.1
XP_016859519.1
15
1765
XM_017004031.1
XP_016859520.1
15
1765
XM_017004032.1
XP_016859521.1
15
1765
XM_017004033.1
XP_016859522.1
15
1765
XM_017004034.1
XP_016859523.1
15
1765
XM_017004035.1
XP_016859524.1
15
1765
XM_017004036.1
XP_016859525.1
15
1765
XM_017004037.1
XP_016859526.1
15
1765
XM_017004038.1
XP_016859527.1
15
1765
XM_017004039.1
XP_016859528.1
15
1765
XM_017004040.1
XP_016859529.1
15
1765
XM_017004041.1
XP_016859530.1
15
1765
XM_017004042.1
XP_016859531.1
15
1765
XM_017004043.1
XP_016859532.1
15
1765
XM_017004044.1
XP_016859533.1
15
1765
XM_017004045.1
XP_016859534.1
15
1765
XM_017004046.1
XP_016859535.1
13
1520
XM_017004047.1
XP_016859536.1
12
1452
XM_017004048.1
XP_016859537.1
11
1379
XM_017004049.1
XP_016859538.1
13
1554
GenBank mRNA
AK307629.1
-
11
976
BX647769.1
-
15
1400
DB516113.1
-
6
251
Gene Symbol: ANKRD36C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001310154.1
NP_001297083.1
17
1925
GenBank mRNA
AK302433.1
-
17
1399
Back To Top
More Information
Gene Ontology Categories:
Process(es)
ion channel inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0