Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lysmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024302.1 | NP_001019473.1 | 2 | 753 | ||
GenBank mRNA | BC089113.1 | - | 2 | 753 | 7319992 | |
BC092612.1 | - | 2 | 115 |