Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM2 SIRNA Assays ›
Gene Symbol:
Tpm2
Gene Name:
tropomyosin 2, beta
Gene Aliases:
-
Chromosome Location:
Chr.5: 59016616 - 59025971 on Build Rnor_6.0
UniGene ID:
Rn.17580
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
257215
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tpm2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001024345.2
NP_001019516.1
2
345
NM_001301235.1
NP_001288164.1
2
345
XM_008763680.1
XP_008761902.1
2
685
XM_008763681.1
XP_008761903.1
2
685
XM_008763682.1
XP_008761904.1
1
63
GenBank mRNA
BC090009.1
-
2
266
7324151
CF110059.1
-
2
231
FQ214609.1
-
2
280
FQ214635.1
-
2
282
FQ214711.1
-
2
282
FQ214816.1
-
2
261
FQ214823.1
-
2
282
FQ214899.1
-
2
282
FQ215016.1
-
2
245
FQ215066.1
-
2
282
FQ215076.1
-
2
242
FQ215143.1
-
2
282
FQ215150.1
-
2
282
FQ215210.1
-
2
282
FQ215302.1
-
2
282
FQ215335.1
-
2
242
FQ215502.1
-
2
282
FQ215601.1
-
2
282
FQ215676.1
-
2
282
FQ215883.1
-
2
280
FQ215920.1
-
2
280
FQ216112.1
-
2
281
FQ216187.1
-
2
282
FQ216228.1
-
2
282
FQ216294.1
-
2
281
FQ216477.1
-
2
282
FQ216483.1
-
2
280
FQ216571.1
-
2
282
FQ216617.1
-
2
284
FQ216760.1
-
2
282
FQ216828.1
-
2
282
FQ216972.1
-
2
283
FQ217401.1
-
2
282
FQ217430.1
-
2
282
FQ217483.1
-
2
285
FQ217946.1
-
2
283
FQ219788.1
-
2
242
FQ219821.1
-
2
282
FQ220371.1
-
2
282
FQ220438.1
-
2
242
FQ220606.1
-
2
282
FQ220675.1
-
2
282
FQ223760.1
-
2
345
FQ224873.1
-
2
284
FQ224893.1
-
2
284
FQ229470.1
-
2
282
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of ATPase activity
actin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0