Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053515.1 | NP_445967.1 | 2 | 593 | ||
GenBank mRNA | BC060533.1 | - | 2 | 597 | 6922056 | |
D12770.1 | - | 2 | 560 | |||
FQ213027.1 | - | 2 | 590 | |||
FQ213086.1 | - | 2 | 590 | |||
FQ213612.1 | - | 2 | 619 | |||
FQ214384.1 | - | 2 | 589 | |||
FQ215448.1 | - | 2 | 593 | |||
FQ217254.1 | - | 2 | 589 | |||
FQ220525.1 | - | 2 | 588 | |||
FQ224814.1 | - | 2 | 590 |