siRNA Details
Gene Symbol: Fdxacb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198675.2 | NP_941077.2 | 4 | 616 | ||
GenBank mRNA | AK035237.1 | - | 2 | 1166 | ||
AK085276.1 | - | 4 | 537 | |||
AK135025.1 | - | 619 | ||||
BC098240.1 | - | 2 | 213 | |||
BC116265.2 | - | 2 | 958 | 40093898 |

More Information
Other Species Targeted

Panther Classification:
