Search
Search

FAM45A| Gene Symbol: FAM45A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303111.1 | NP_001290040.1 | 10 | 2220 | ||
| NM_001303112.1 | NP_001290041.1 | 9 | 1967 | |||
| NM_001303113.1 | NP_001290042.1 | 8 | 1886 | |||
| NM_207009.3 | NP_996892.1 | 9 | 2015 | |||
| NR_130122.1 | 8 | 1903 | ||||
| XM_006717849.3 | XP_006717912.1 | 9 | 2035 | |||
| XM_006717850.3 | XP_006717913.1 | 10 | 2277 | |||
| XM_006717851.3 | XP_006717914.1 | 10 | 2257 | |||
| XM_006717852.3 | XP_006717915.1 | 10 | 2241 | |||
| XM_017016261.1 | XP_016871750.1 | 10 | 2750 | |||
| XM_017016262.1 | XP_016871751.1 | 11 | 2465 | |||
| XM_017016263.1 | XP_016871752.1 | 11 | 2446 | |||
| XM_017016264.1 | XP_016871753.1 | 9 | 1410 | |||
| XM_017016265.1 | XP_016871754.1 | 10 | 2261 | |||
| XM_017016266.1 | XP_016871755.1 | 11 | 2442 | |||
| XM_017016267.1 | XP_016871756.1 | 11 | 2426 | |||
| GenBank mRNA | AK027029.1 | - | 7 | 2115 | ||