Search
Search

ZNF320| Gene Symbol: ZNF320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207333.2 | NP_997216.2 | 4 | 6003 | ||
| XM_006723059.3 | XP_006723122.1 | 5 | 6049 | |||
| XM_017026393.1 | XP_016881882.1 | 6 | 6294 | |||
| XM_017026394.1 | XP_016881883.1 | 6 | 6219 | |||
| XM_017026395.1 | XP_016881884.1 | 6 | 6367 | |||
| XM_017026396.1 | XP_016881885.1 | 5 | 6136 | |||
| XM_017026397.1 | XP_016881886.1 | 5 | 6141 | |||
| XM_017026398.1 | XP_016881887.1 | 5 | 6020 | |||
| XM_017026399.1 | XP_016881888.1 | 4 | 6013 | |||
| XM_017026400.1 | XP_016881889.1 | 4 | 5933 | |||
| GenBank mRNA | AF086262.1 | - | 1 | 290 | ||
| AK094328.1 | - | 1 | 1720 | |||
| AK095630.1 | - | 2 | 3109 | |||