Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013842.2 | NP_001013864.1 | 7 | 1497 | ||
NM_001198827.1 | NP_001185756.1 | 6 | 1390 | |||
NM_173686.2 | NP_775957.2 | 7 | 1473 | |||
GenBank mRNA | AK092034.1 | - | 6 | 1281 | ||
BC012750.2 | - | 6 | 1289 | 3618901 |