Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERMARD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278531.1 | NP_001265460.1 | 2 | 65 | ||
NM_001278533.1 | NP_001265462.1 | 2 | 65 | |||
NM_018341.2 | NP_060811.1 | 2 | 65 | |||
XM_011535938.2 | XP_011534240.1 | 2 | 65 | |||
XM_011535939.2 | XP_011534241.1 | 2 | 1155 | |||
XM_011535940.2 | XP_011534242.1 | 2 | 964 | |||
XM_017011027.1 | XP_016866516.1 | 2 | 964 | |||
XM_017011028.1 | XP_016866517.1 | 2 | 289 | |||
XM_017011029.1 | XP_016866518.1 | 2 | 49 | |||
XM_017011032.1 | XP_016866521.1 | 2 | 1155 | |||
XM_017011033.1 | XP_016866522.1 | 2 | 964 | |||
GenBank mRNA | AK002014.1 | - | 2 | 62 | ||
BC044812.1 | - | 2 | 49 | |||
DA650477.1 | - | 2 | 65 |