Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SPIRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128626.1 | NP_001122098.1 | 17 | 5013 | ||
NM_001128627.1 | NP_001122099.1 | 16 | 4669 | |||
NM_020148.2 | NP_064533.3 | 16 | 4971 | |||
XM_005258122.4 | XP_005258179.1 | 17 | 5328 | |||
XM_011525701.1 | XP_011524003.1 | 18 | 5376 | |||
XM_011525702.1 | XP_011524004.1 | 18 | 4888 | |||
GenBank mRNA | AB032961.2 | - | 17 | 5065 | ||
AJ277587.1 | - | 14 | 4560 |