Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM118A SIRNA Assays ›
Gene Symbol:
FAM118A
Gene Name:
family with sequence similarity 118 member A
Gene Aliases:
C22orf8
Chromosome Location:
Chr.22: 45309200 - 45341955 on Build GRCh38
UniGene ID:
Hs.265018
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
25835
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM118A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001104595.1
NP_001098065.1
4
860
NM_017911.2
NP_060381.2
3
533
XM_011530247.2
XP_011528549.1
3
637
XM_011530254.2
XP_011528556.1
4
556
XM_011530256.1
XP_011528558.1
3
425
XM_017028841.1
XP_016884330.1
3
422
XM_017028842.1
XP_016884331.1
3
425
XM_017028843.1
XP_016884332.1
3
422
XM_017028844.1
XP_016884333.1
3
425
XM_017028845.1
XP_016884334.1
4
622
XM_017028846.1
XP_016884335.1
3
506
XM_017028847.1
XP_016884336.1
3
1150
XM_017028848.1
XP_016884337.1
3
548
XM_017028849.1
XP_016884338.1
3
384
XM_017028850.1
XP_016884339.1
3
507
XM_017028851.1
XP_016884340.1
3
1138
XM_017028852.1
XP_016884341.1
3
544
XM_017028853.1
XP_016884342.1
3
425
XM_017028854.1
XP_016884343.1
3
507
XM_017028855.1
XP_016884344.1
4
557
XM_017028856.1
XP_016884345.1
4
557
XM_017028857.1
XP_016884346.1
3
506
XM_017028858.1
XP_016884347.1
3
507
XM_017028859.1
XP_016884348.1
3
506
XM_017028860.1
XP_016884349.1
3
507
XM_017028861.1
XP_016884350.1
3
507
GenBank mRNA
AK000642.1
-
3
503
AK125027.1
-
4
1078
BC013696.1
-
3
290
3860694
CR456403.1
-
2
259
DB153788.1
-
3
319
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-8nwch:80/10.239.162.255:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0