Search
Search

Lrrn4cl| Gene Symbol: Lrrn4cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013019.2 | NP_001013037.1 | 2 | 402 | ||
| GenBank mRNA | AK087615.1 | - | 403 | |||
| AK158268.1 | - | 2 | 340 | |||
| BC107244.2 | - | 2 | 375 | 40052213 | ||
| BC107245.2 | - | 2 | 375 | 40052216 | ||