Search
Search

Ttll4| Gene Symbol: Ttll4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014974.1 | NP_001014974.1 | 19, 20 | 3719 | ||
| GenBank mRNA | AK129075.1 | - | 17, 18 | 2546 | ||
| AM690747.1 | - | 17, 18 | 3322 | |||
| BC044790.1 | - | 6, 7 | 696 | |||
| BC049820.1 | - | 4, 5 | 950 | |||
| BC085151.1 | - | 19, 20 | 3719 | 30534216 | ||