Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008105.3 | NP_032131.2 | 3 | 4254 | ||
NM_023887.4 | NP_076376.3 | 3 | 4232 | |||
NM_133219.1 | NP_573482.1 | 3 | 3894 | |||
GenBank mRNA | AK077598.1 | - | 3 | 3871 | ||
AK080061.1 | - | 3 | 3823 | |||
AK146638.1 | - | 3 | 2996 | |||
AK146679.1 | - | 3 | 2997 | |||
AK156328.1 | - | 3 | 3761 | |||
AK169167.1 | - | 4 | 3042 | |||
AY435148.1 | - | 3 | 3645 | |||
AY435149.1 | - | 3 | 3645 | |||
AY435150.1 | - | 3 | 3645 | |||
BC054845.1 | - | 3 | 3876 | 6493137 | ||
BC094572.1 | - | 3 | 4233 | 30356538 | ||
BM933481.1 | - | 1 | 261 |