Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027660.1 | NP_081936.1 | 2 | 329 | ||
GenBank mRNA | AK016718.1 | - | 2 | 330 | ||
AY252101.1 | - | 2 | 329 | |||
BC127605.1 | - | 2 | 227 | 40057157 | ||
BC128270.1 | - | 2 | 227 | 40057161 | ||
BC139074.1 | - | 2 | 296 | 8862096 | ||
BC139075.1 | - | 2 | 296 | 8862097 |