siRNA Details
Gene Symbol: SH3PXD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017995.2 | NP_001017995.1 | 13 | 6485 | ||
XM_017009351.1 | XP_016864840.1 | 14 | 6569 | |||
GenBank mRNA | AB037716.1 | - | 2 | 5235 | ||
AK000838.1 | - | 1 | 340 | |||
AK095834.1 | - | 1 | 1367 | |||
BC016513.2 | - | 1 | 493 | |||
BC065230.1 | - | 1 | 253 | |||
BC110316.1 | - | 1 | 677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
