Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MACF1 SIRNA Assays ›
Gene Symbol:
MACF1
Gene Name:
microtubule-actin crosslinking factor 1
Gene Aliases:
ABP620, ACF7, MACF, OFC4
Chromosome Location:
Chr.1: 39084167 - 39487138 on Build GRCh38
UniGene ID:
Hs.472475
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
258848
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MACF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012090.5
NP_036222.3
52
8616
XM_005270694.1
XP_005270751.1
53
15478
XM_005270695.1
XP_005270752.1
53
15478
XM_005270696.1
XP_005270753.1
53
15478
XM_005270697.1
XP_005270754.1
53
15478
XM_005270698.1
XP_005270755.1
53
15478
XM_006710519.1
XP_006710582.1
53
15478
XM_006710520.2
XP_006710583.1
52
15415
XM_006710521.1
XP_006710584.1
53
15478
XM_006710522.1
XP_006710585.1
53
15478
XM_006710524.2
XP_006710587.1
52
15244
XM_006710526.3
XP_006710589.1
58
14910
XM_006710528.3
XP_006710591.1
58
15471
XM_006710529.3
XP_006710592.1
58
14828
XM_006710530.3
XP_006710593.1
58
15716
XM_006710531.3
XP_006710594.1
58
15447
XM_006710532.3
XP_006710595.1
57
15375
XM_006710533.2
XP_006710596.1
57
15375
XM_006710535.3
XP_006710598.1
58
14980
XM_006710536.2
XP_006710599.1
48
9277
XM_011541141.2
XP_011539443.1
53
15424
XM_011541142.2
XP_011539444.1
52
10027
XM_017000873.1
XP_016856362.1
53
15478
XM_017000874.1
XP_016856363.1
53
15478
XM_017000875.1
XP_016856364.1
57
14800
XM_017000876.1
XP_016856365.1
57
14884
XM_017000877.1
XP_016856366.1
58
14806
XM_017000878.1
XP_016856367.1
58
14669
XM_017000879.1
XP_016856368.1
23
10240
XM_017000880.1
XP_016856369.1
47
9214
XM_017000881.1
XP_016856370.1
52
8591
XM_017000882.1
XP_016856371.1
47
9214
GenBank mRNA
AB007934.3
-
31
5901
AB029290.1
-
52
8615
AF317696.1
-
22
10190
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of epithelial cell migration
Wnt signaling pathway
positive regulation of Wnt signaling pathway
regulation of microtubule-based process
wound healing
Golgi to plasma membrane protein transport
positive regulation of axon extension
regulation of focal adhesion assembly
cell-cell adhesion
actin binding
calcium ion binding
protein binding
microtubule binding
ATPase activity
poly(A) RNA binding
actin filament binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-wfnwd:80/10.239.165.170:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0