Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001734.1 | NP_001001734.1 | 5 | 600 | ||
NM_144699.3 | NP_653300.2 | 22 | 3561 | |||
XM_011509582.1 | XP_011507884.1 | 21 | 3028 | |||
GenBank mRNA | AF506797.1 | - | 22 | 3553 | ||
AL833335.1 | - | 22 | 3534 | |||
BC028297.1 | - | 5 | 600 | |||
BC094801.1 | - | 22 | 3512 | 5268907 | ||
BX647234.1 | - | 22 | 3535 |