Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152641.2 | NP_689854.2 | 21 | 5555 | ||
XM_006719272.3 | XP_006719335.1 | 22 | 5766 | |||
XM_011538025.1 | XP_011536327.1 | 15 | 4662 | |||
GenBank mRNA | AB046777.1 | - | 7 | 2468 | ||
AK027617.1 | - | 7 | 1190 | |||
AK075385.1 | - | 7 | 2109 | |||
AK127872.1 | - | 12 | 3376 | |||
AL832200.1 | - | 17 | 5108 | |||
BC018661.1 | - | 6 | 763 | |||
BC038726.1 | - | 7 | 856 | |||
BC090062.1 | - | 7 | 2630 | |||
BX537879.1 | - | 7 | 3183 | |||
BX648152.1 | - | 15 | 4843 | |||
BX649133.1 | - | 13 | 4401 |