Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008865.1 | NP_001008865.1 | 2 | 217 | ||
XM_006243993.2 | XP_006244055.1 | 3 | 457 |