Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRAK1BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010844.3 | NP_001010844.1 | 1 | 318 | ||
XM_006715336.3 | XP_006715399.1 | 1 | 359 | |||
XM_011535447.1 | XP_011533749.1 | 1 | 358 | |||
XM_011535448.2 | XP_011533750.1 | 1 | 376 | |||
XM_017010265.1 | XP_016865754.1 | 1 | 376 | |||
GenBank mRNA | AK309003.1 | - | 1 | 255 | ||
BC112253.1 | - | 1 | 307 | 8327721 | ||
BC112255.1 | - | 1 | 307 | 8327723 | ||
CR990080.1 | - | 1 | 318 |