Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012047.1 | NP_001012047.1 | 4 | 1673 | ||
XM_006252609.2 | XP_006252671.1 | 6 | 1808 | |||
XM_006252610.1 | XP_006252672.1 | 5 | 1693 | |||
XM_006252611.2 | XP_006252673.1 | 5 | 2887 | |||
XM_006252612.1 | XP_006252674.1 | 7 | 2198 | |||
XM_006252614.1 | XP_006252676.1 | 6 | 2075 | |||
XM_006252615.2 | XP_006252677.1 | 5 | 2023 | |||
XM_006252616.2 | XP_006252678.1 | 5 | 2804 | |||
XM_006252617.1 | XP_006252679.1 | 5 | 1927 | |||
XM_006252618.1 | XP_006252680.1 | 4 | 1660 | |||
XM_006252619.2 | XP_006252681.1 | 7 | 1835 | |||
XM_006252620.2 | XP_006252682.1 | 6 | 1849 | |||
XM_006252622.2 | XP_006252684.1 | 5 | 1804 | |||
XM_008770998.1 | XP_008769220.1 | 5 | 1647 | |||
XM_008770999.1 | XP_008769221.1 | 5 | 1964 | |||
GenBank mRNA | BC090017.1 | - | 4 | 1673 | 7325227 |