Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012119.1 | NP_001012119.1 | 5 | 1984 | ||
XM_006241994.2 | XP_006242056.1 | 5 | 2151 | |||
GenBank mRNA | BC087122.1 | - | 5 | 1984 | 7108347 |