Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Terf2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013143.1 | NP_001013161.1 | 1 | 486 | ||
GenBank mRNA | BC090335.1 | - | 1 | 486 | 7302641 |