Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcgf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013154.1 | NP_001013172.1 | 9 | 1693 | ||
XM_006231509.2 | XP_006231571.1 | 10 | 1757 | |||
XM_008760437.1 | XP_008758659.1 | 9 | 1764 | |||
XM_008760438.1 | XP_008758660.1 | 10 | 1909 | |||
GenBank mRNA | BC083820.1 | - | 9 | 1693 | 7111130 |