Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXorf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184782.1 | NP_001171711.1 | 3 | 973 | ||
NM_018015.5 | NP_060485.4 | 3 | 973 | |||
GenBank mRNA | AK001040.1 | - | 3 | 928 | ||
AK024253.1 | - | 3 | 247 | |||
AK291730.1 | - | 3 | 922 | |||
BC011483.1 | - | 3 | 894 | 3453700 | ||
BC033709.1 | - | 3 | 885 | 3910777 | ||
BC054032.2 | - | 3 | 861 | |||
BC070110.1 | - | 3 | 924 | 5264812 |