Search
Search

BSDC1| Gene Symbol: BSDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143888.2 | NP_001137360.1 | 4 | 311 | ||
| NM_001300958.1 | NP_001287887.1 | 4 | 311 | |||
| NM_018045.7 | NP_060515.3 | 4 | 260 | |||
| NR_125352.1 | 4 | 260 | ||||
| XM_005270986.4 | XP_005271043.1 | 4 | 240 | |||
| XM_011541677.2 | XP_011539979.1 | 4 | 558 | |||
| GenBank mRNA | AK001138.1 | - | 4 | 230 | ||
| AK021440.1 | - | 4 | 239 | |||
| AK225026.1 | - | 4 | 241 | |||
| AK225717.1 | - | 4 | 230 | |||
| AK297352.1 | - | 4 | 239 | |||
| AK300306.1 | - | 4 | 280 | |||
| BC010573.1 | - | 4 | 219 | |||
| BC026322.1 | - | 3 | 227 | 4794111 | ||
| BC028381.2 | - | 4 | 232 | 4812029 | ||
| BP363955.1 | - | 4 | 260 | |||
| BX641056.1 | - | 3 | 277 | |||
| CR457252.1 | - | 3 | 213 | |||