Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014996.2 | NP_001014996.2 | 7 | 1583 | ||
GenBank mRNA | AK077045.1 | - | 7 | 1583 | ||
BC092399.1 | - | 7 | 1569 | 30549055 |