Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024781.1 | NP_001019952.1 | 2 | 1067 | ||
GenBank mRNA | BC097973.1 | - | 2 | 1067 | 7442067 |