Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024792.1 | NP_001019963.1 | 5 | 403 | ||
XM_006247374.2 | XP_006247436.1 | 5 | 741 | |||
XM_006247376.2 | XP_006247438.1 | 5 | 426 | |||
GenBank mRNA | BC081862.1 | - | 5 | 429 | ||
BC097349.1 | - | 5 | 403 | 7460172 | ||
FQ228904.1 | - | 5 | 393 |