Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MUC1 SIRNA Assays ›
          Gene Symbol:
          MUC1
          Gene Name:
          mucin 1, cell surface associated
          Gene Aliases:
          ADMCKD, ADMCKD1, CA 15-3, CD227, EMA, H23AG, KL-6, MAM6, MCD, MCKD, MCKD1, MUC-1, MUC-1/SEC, MUC-1/X, MUC1/ZD, PEM, PEMT, PUM
          Chromosome Location:
          Chr.1: 155185824 - 155192915 on Build GRCh38
          UniGene ID:
          Hs.89603
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 261005

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MUC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001018016.2 NP_001018016.1 4, 5 379
          NM_001018017.2 NP_001018017.1 4, 5 352
          NM_001204285.1 NP_001191214.1 4, 5 1012
          NM_001204286.1 NP_001191215.1 4, 5 1039
          NM_001204287.1 NP_001191216.1 4, 5 433
          NM_001204288.1 NP_001191217.1 4, 5 379
          NM_001204289.1 NP_001191218.1 3, 4 301
          NM_001204290.1 NP_001191219.1 3, 4 238
          NM_001204291.1 NP_001191220.1 3, 4 310
          NM_001204293.1 NP_001191222.1 4, 5 406
          NM_002456.5 NP_002447.4 4, 5 406
          GenBank mRNA AF125525.1 - 4, 5 271
          AK301575.1 - 5, 6 1264
          AY327587.1 - 4, 5 357
          AY327588.1 - 3, 4 252
          AY327596.1 - 5, 6 487
          AY466157.1 - 4, 5 261
          BC120974.2 - 4, 5 329 40115473
          BC120975.2 - 4, 5 356 40115475
          EF583642.1 - 4, 5 682
          EF583643.1 - 4, 5 510
          EF583644.1 - 4, 5 496
          EF583645.1 - 4, 5 456
          EF583646.1 - 4, 5 424
          EF583647.1 - 4, 5 411
          EF583648.1 - 4, 5 338
          EF583649.1 - 4, 5 357
          EF583650.1 - 4, 5 294
          EF583652.1 - 3, 4 279
          EF583654.1 - 3, 4 216
          EF670706.1 - 4, 5 706
          EF670707.1 - 4, 5 375
          EF670708.1 - 3, 4 288
          EF670711.1 - 4, 5 384
          FJ226028.1 - 3, 4 813
          FJ226029.1 - 3, 4 971
          FJ226030.1 - 4, 5 941
          FJ226031.1 - 3, 4 899
          FJ226032.1 - 3, 4 832
          FJ226033.1 - 3, 4 813
          FJ226034.1 - 4, 5 732
          FJ226037.1 - 4, 5 510
          FJ226039.1 - 4, 5 437
          FJ226040.1 - 4, 5 357
          FJ226041.1 - 5, 6 514
          FJ226042.1 - 4, 5 411
          FJ226043.1 - 4, 5 397
          FJ226045.1 - 3, 4 351
          FJ226047.1 - 4, 5 311
          FJ226052.1 - 4, 5 330
          J05581.1 - 4, 5 1012
          J05582.1 - 3353
          M34088.1 - 2, 3 499
          M34089.1 - 2, 3 499
          U60260.1 - 4, 5 305
          U60261.1 - 4, 5 359
          X52229.1 - 5, 6 1117
          X80761.1 - 4, 5 280

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
          negative regulation of transcription by competitive promoter binding
          O-glycan processing
          negative regulation of cell adhesion mediated by integrin
          positive regulation of transcription from RNA polymerase II promoter in response to stress
          regulation of transcription from RNA polymerase II promoter in response to stress
          positive regulation of histone H4 acetylation
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          p53 binding
          transcription cofactor activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0