Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013602.3 | NP_038630.1 | 3 | 521 | ||
GenBank mRNA | AA050166.1 | - | 3 | 300 | ||
AK007983.1 | - | 3 | 370 | |||
AK011040.1 | - | 3 | 370 | |||
AK018727.1 | - | 3 | 384 | |||
AK154954.1 | - | 1 | 1067 | |||
BC036990.1 | - | 366 | 4990006 |