Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SRGAP2B SIRNA Assays ›
          Gene Symbol:
          SRGAP2B SRGAP2 SRGAP2C SRGAP2D
          Gene Name:
          SLIT-ROBO Rho GTPase activating protein 2B
          SLIT-ROBO Rho GTPase activating protein 2
          SLIT-ROBO Rho GTPase activating protein 2C
          SLIT-ROBO Rho GTPase activating protein 2D (pseudogene)
          Gene Aliases:
          SRGAP2L, SRGAP2P2
          ARHGAP34, FNBP2, SRGAP2A, SRGAP3
          SRGAP2P1
          -
          Chromosome Location:
          Chr.1: 144887191 - 145093441 on Build GRCh38
          Chromosome Location:
          Chr.1: 206203464 - 206464443 on Build GRCh38
          Chromosome Location:
          Chr.1: 121184967 - 121392819 on Build GRCh38
          Chromosome Location:
          Chr.1: 143972639 - 144069704 on Build GRCh38
          UniGene ID:
          Hs.497575
          Hs.497575
          Hs.497575
          Hs.497575
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID 261179

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SRGAP2B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271870.2 NP_001258799.2 5 1039
          XM_005277421.3 XP_005277478.1 3 762
          XM_005277422.4 XP_005277479.1 3 176
          XM_017002085.1 XP_016857574.1 5 1118
          XM_017002086.1 XP_016857575.1 5 964
          XM_017002087.1 XP_016857576.1 5 1118
          XM_017002088.1 XP_016857577.1 6 1179
          XM_017002089.1 XP_016857578.1 7 1227
          XM_017002090.1 XP_016857579.1 6 1179
          XM_017002091.1 XP_016857580.1 7 1227
          XM_017002092.1 XP_016857581.1 3 762
          GenBank mRNA BQ706477.1 - 1 93
          BY795901.2 - 3 186
          HY154277.1 - 4 366
          Gene Symbol: SRGAP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001170637.3 NP_001164108.1 5 1120
          NM_001300952.1 NP_001287881.1 5 1120
          NM_015326.4 NP_056141.2 5 1120
          XM_005277510.2 XP_005277567.1 6 1307
          XM_005277511.2 XP_005277568.1 6 1307
          XM_005277514.1 XP_005277571.1 6 1307
          XM_005277515.1 XP_005277572.1 6 1307
          XM_011509354.1 XP_011507656.1 6 1307
          XM_011509355.1 XP_011507657.1 6 1307
          XM_011509356.2 XP_011507658.1 6 1307
          XM_011509359.1 XP_011507661.1 6 1307
          XM_017000838.1 XP_016856327.1 4 951
          XM_017000839.1 XP_016856328.1 4 981
          XM_017000840.1 XP_016856329.1 6 596
          XM_017000841.1 XP_016856330.1 6 1307
          GenBank mRNA AB007925.1 - 5 650
          AK293335.1 - 2 156
          AK295845.1 - 5 876
          BC132872.1 - 5 648 40146894
          BC132874.1 - 5 648 40146896
          BC144343.1 - 5 648 9052863
          S68466.1 - 2 138
          Gene Symbol: SRGAP2C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271872.1 NP_001258801.1 6 1183
          XM_005277497.3 XP_005277554.1 6 1278
          XM_005277499.4 XP_005277556.1 3 177
          XM_011542015.2 XP_011540317.1 5 751
          XM_017002149.1 XP_016857638.1 4 922
          GenBank mRNA BC112927.1 - 6 1183
          Gene Symbol: SRGAP2D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_120535.1 4 893

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          nervous system development
          lamellipodium assembly involved in ameboidal cell migration
          signal transduction
          cell proliferation
          extension of a leading process involved in cell motility in cerebral cortex radial glia guided migration
          substrate adhesion-dependent cell spreading
          positive regulation of GTPase activity
          filopodium assembly
          neuron projection morphogenesis
          actin filament severing
          regulation of small GTPase mediated signal transduction
          negative regulation of cell death
          dendritic spine development
          negative regulation of neuron migration
          negative regulation of cell migration
          negative regulation of filopodium assembly
          negative regulation of dendritic spine development
          positive regulation of neuron migration
          GTPase activator activity
          protein binding
          protein homodimerization activity
          Rac GTPase binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0