Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNT3 SIRNA Assays ›
Gene Symbol:
Tnnt3
Gene Name:
troponin T type 3 (skeletal, fast)
Gene Aliases:
Tnt
Chromosome Location:
Chr.1: 215666628 - 215683628 on Build Rnor_6.0
UniGene ID:
Rn.15488
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
261429
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tnnt3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270664.1
NP_001257593.1
8
245
NM_001270665.1
NP_001257594.1
10
278
NM_001270666.1
NP_001257595.1
10
278
NM_001270668.1
NP_001257597.1
8
245
NM_001270670.1
NP_001257599.1
9
260
NM_001270671.1
NP_001257600.1
9
260
NM_001270672.1
NP_001257601.1
8
251
NM_001270673.1
NP_001257602.1
9
263
NM_001270675.1
NP_001257604.1
8
251
NM_001270676.1
NP_001257605.1
9
263
NM_001270677.1
NP_001257606.1
7
233
NM_001270678.1
NP_001257607.1
9
266
NM_031532.2
NP_113720.1
9
266
XM_006230681.2
XP_006230743.1
8
208
XM_006230683.2
XP_006230745.1
7
196
XM_006230685.2
XP_006230747.1
6
178
XM_006230686.2
XP_006230748.1
6
178
XM_008760080.1
XP_008758302.1
11
312
XM_008760081.1
XP_008758303.1
11
312
XM_008760082.1
XP_008758304.1
10
262
XM_008760083.1
XP_008758305.1
10
259
XM_008760084.1
XP_008758306.1
9
285
XM_008760085.1
XP_008758307.1
9
250
XM_008760086.1
XP_008758308.1
8
232
GenBank mRNA
CB612473.1
-
10
227
DQ062204.1
-
9
162
DQ273678.1
-
9
162
FQ214650.1
-
9
226
FQ214724.1
-
9
227
FQ214766.1
-
8
207
FQ214833.1
-
9
228
FQ215030.1
-
8
207
FQ215059.1
-
8
238
FQ215106.1
-
8
207
FQ215114.1
-
8
213
FQ215252.1
-
9
225
FQ215316.1
-
8
227
FQ215319.1
-
8
207
FQ215327.1
-
10
240
FQ215374.1
-
9
253
FQ215598.1
-
10
237
FQ215602.1
-
8
240
FQ215694.1
-
8
245
FQ215723.1
-
10
279
FQ215881.1
-
10
240
FQ215885.1
-
10
228
FQ215897.1
-
9
253
FQ215955.1
-
8
207
FQ216024.1
-
5
207
FQ216059.1
-
10
235
FQ216097.1
-
9
221
FQ216131.1
-
8
238
FQ216208.1
-
9
225
FQ216324.1
-
8
207
FQ216367.1
-
8
207
FQ216417.1
-
9
226
FQ216467.1
-
8
207
FQ216480.1
-
8
213
FQ216535.1
-
9
222
FQ216561.1
-
5
207
FQ216694.1
-
9
222
FQ216711.1
-
8
244
FQ216750.1
-
5
98
FQ216763.1
-
6
198
FQ216784.1
-
8
213
FQ216866.1
-
10
240
FQ217147.1
-
8
206
FQ217846.1
-
10
272
FQ217900.1
-
9
253
FQ223840.1
-
8
207
FQ223999.1
-
8
207
FQ224006.1
-
8
209
FQ224794.1
-
8
207
FQ224813.1
-
10
240
FQ224836.1
-
8
207
FQ224839.1
-
9
225
FQ224892.1
-
9
228
FQ224966.1
-
9
211
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
muscle contraction
regulation of muscle contraction
regulation of striated muscle contraction
metabolic process
regulation of ATPase activity
actin binding
tropomyosin binding
troponin C binding
calcium-dependent ATPase activity
troponin I binding
troponin T binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0