Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317056.1 | NP_001303985.1 | 9 | 2248 | ||
NR_073169.1 | 9 | 1100 | ||||
NR_133652.1 | 9 | 2248 | ||||
XM_011516065.1 | XP_011514367.1 | 9 | 2248 | |||
XM_011516066.2 | XP_011514368.1 | 9 | 2248 | |||
GenBank mRNA | AY316116.1 | - | 4 | 586 | ||
AY515311.1 | - | 9 | 2248 | |||
BC128587.1 | - | 9 | 1066 | 40129083 | ||
BX420361.2 | - | 4 | 681 |