Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CADM1 SIRNA Assays ›
          Gene Symbol:
          Cadm1
          Gene Name:
          cell adhesion molecule 1
          Gene Aliases:
          2900073G06Rik, 3100001I08Rik, AI987920, Bl2, Igsf4, Igsf4a, Necl2, RA175, RA175A, RA175B, RA175C, RA175N, ST17, SgIGSF, SynCam, Tslc1
          Chromosome Location:
          Chr.9: 47530239 - 47853385 on Build GRCm38
          UniGene ID:
          Mm.234832
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 261715

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cadm1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001025600.1 NP_001020771.1 7 1001
          NM_018770.3 NP_061240.3 7 1001
          NM_207675.2 NP_997558.2 7 1001
          NM_207676.2 NP_997559.1 7 1001
          GenBank mRNA AB052293.1 - 7 1001
          AB064265.2 - 7 980
          AB092414.1 - 7 980
          AB183399.1 - 7 980
          AB183400.1 - 7 980
          AB183401.1 - 7 980
          AB183402.1 - 7 980
          AB377192.1 - 7 1001
          AF061260.1 - 6 838
          AF434663.1 - 7 1096
          AF539424.1 - 7 993
          AK013775.1 - 7 1002
          AK013911.1 - 7 1095
          AK053839.1 - 7 1002
          AK207220.1 - 1 38
          AK210200.1 - 1 38
          AY351388.1 - 7 980
          BC057125.1 - 7 989 6414884
          BC090970.1 - 7 1180 6703838
          BC094661.1 - 4 551
          BC095986.1 - 7 993 5372277
          BQ043739.1 - 1 15
          DQ279856.1 - 7 980
          EU541474.1 - 1 150
          EU541475.1 - 1 150
          EU541476.1 - 1 150
          EU541477.1 - 1 150
          EU541478.1 - 1 150
          JQ782219.1 - 7 980

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          immune system process
          apoptotic process
          cell adhesion
          homophilic cell adhesion via plasma membrane adhesion molecules
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          multicellular organism development
          spermatogenesis
          synapse assembly
          cell recognition
          unidimensional cell growth
          calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules
          cell differentiation
          susceptibility to natural killer cell mediated cytotoxicity
          positive regulation of natural killer cell mediated cytotoxicity
          positive regulation of cytokine secretion
          detection of stimulus
          bone development
          receptor binding
          protein binding
          PDZ domain binding
          protein homodimerization activity
          cell adhesion molecule binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0