Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BCL2L11 SIRNA Assays ›
          Gene Symbol:
          Bcl2l11
          Gene Name:
          BCL2-like 11 (apoptosis facilitator)
          Gene Aliases:
          1500006F24Rik, Bim, Bod, bcl2-L-11
          Chromosome Location:
          Chr.2: 128126038 - 128162547 on Build GRCm38
          UniGene ID:
          Mm.141083
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 261717

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Bcl2l11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009754.3 NP_033884.1 4 4167
          NM_207680.2 NP_997563.1 4 4425
          NM_207681.2 NP_997564.1 5 4257
          GenBank mRNA AA275665.1 - 1 376
          AA881651.1 - 1 403
          CA321725.1 - 1 548

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_171988, XM_006234983, XM_006234985, XM_006234987.

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          leukocyte homeostasis
          B cell homeostasis
          B cell apoptotic process
          kidney development
          lymphocyte homeostasis
          myeloid cell homeostasis
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cell-matrix adhesion
          spermatogenesis
          male gonad development
          intrinsic apoptotic signaling pathway in response to DNA damage
          post-embryonic development
          positive regulation of cell death
          mammary gland development
          positive regulation of protein homooligomerization
          response to endoplasmic reticulum stress
          tube formation
          odontogenesis of dentin-containing tooth
          regulation of apoptotic process
          T cell homeostasis
          positive regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of neuron apoptotic process
          ear development
          positive regulation of cell cycle
          regulation of organ growth
          developmental pigmentation
          regulation of developmental pigmentation
          spleen development
          thymus development
          post-embryonic organ morphogenesis
          positive regulation of apoptotic process by virus
          cellular process regulating host cell cycle in response to virus
          thymocyte apoptotic process
          positive regulation of release of cytochrome c from mitochondria
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of mitochondrial membrane permeability involved in apoptotic process
          apoptotic process involved in embryonic digit morphogenesis
          positive regulation of fibroblast apoptotic process
          positive regulation of intrinsic apoptotic signaling pathway
          protein binding
          microtubule binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0