Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xkr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012229.1 | NP_001012229.1 | 2 | 584 | ||
XM_006237733.2 | XP_006237795.1 | 2 | 667 | |||
GenBank mRNA | AY534263.1 | - | 2 | 584 |