Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033606.2 | NP_001028778.2 | 3 | 858 | ||
NM_001136222.1 | NP_001129694.1 | 2 | 440 | |||
NM_028817.3 | NP_083093.2 | 4 | 998 | |||
GenBank mRNA | AK012088.1 | - | 3 | 801 | ||
AK160839.1 | - | 2 | 384 | |||
AK161708.1 | - | 4 | 999 | |||
AK166723.1 | - | 4 | 936 | |||
AK195074.1 | - | 2 | 218 | |||
BC031529.1 | - | 2 | 583 | 4009004 |