Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSEN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000447.2 | NP_000438.2 | 8 | 1122 | ||
NM_012486.2 | NP_036618.2 | 8 | 1122 | |||
XM_005273199.3 | XP_005273256.1 | 7 | 935 | |||
XM_017001835.1 | XP_016857324.1 | 7 | 1443 | |||
XM_017001836.1 | XP_016857325.1 | 7 | 935 | |||
GenBank mRNA | AF416718.1 | - | 6 | 763 | ||
AK292299.1 | - | 9 | 1074 | |||
AK301143.1 | - | 7 | 871 | |||
BC006365.2 | - | 9 | 1316 | 4131408 | ||
BT006984.1 | - | 6 | 695 | |||
L43964.1 | - | 7 | 1062 | |||
L44577.1 | - | 8 | 1060 | |||
U34349.1 | - | 6 | 786 |